siRNA Details
Gene Symbol: Snx13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108708.1 | NP_001102178.1 | 18, 19 | 2212 | ||
XM_006240022.2 | XP_006240084.1 | 21, 22 | 2934 | |||
XM_006240023.2 | XP_006240085.1 | 21, 22 | 2899 |
