siRNA Details
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108718.1 | NP_001102188.1 | 5 | 609 | ||
XM_006240394.2 | XP_006240456.1 | 5 | 670 | |||
GenBank mRNA | FQ225419.1 | - | 3 | 245 | ||
FQ226790.1 | - | 2 | 176 | |||
FQ230972.1 | - | 3 | 245 | |||
FQ232964.1 | - | 4 | 285 | |||
FQ234582.1 | - | 2 | 176 |
