siRNA Details
Gene Symbol: Gskip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014198.2 | NP_001014220.2 | 3 | 406 | ||
XM_006240527.2 | XP_006240589.1 | 3 | 777 | |||
GenBank mRNA | BC087681.1 | - | 3 | 383 | 7321205 | |
FM111441.1 | - | 3 | 409 |
