siRNA Details
Gene Symbol: Nabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034939.1 | NP_001030111.2 | 2, 3 | 624 | ||
NM_001244819.1 | NP_001231748.1 | 3, 4 | 347 | |||
XM_006240769.2 | XP_006240831.1 | 3, 4 | 588 | |||
XM_006240770.2 | XP_006240832.1 | 3, 4 | 443 | |||
XM_006240771.2 | XP_006240833.1 | 3, 4 | 438 | |||
XM_008765029.1 | XP_008763251.1 | 3, 4 | 603 | |||
GenBank mRNA | BC104710.1 | - | 2, 3 | 624 | 7319804 | |
DV214819.1 | - | 3, 4 | 347 | |||
FQ219805.1 | - | 3, 4 | 335 |
