siRNA Details
Gene Symbol: Cdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199501.1 | NP_955795.1 | 7 | 1009 | ||
XM_006240778.1 | XP_006240840.1 | 8 | 1229 | |||
GenBank mRNA | BC061832.1 | - | 7 | 1009 | 5599608 | |
D28753.1 | - | 8 | 1045 |
