siRNA Details
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240779.2 | XP_006240841.1 | 10 | 2251 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021882, BC082555, AK131956, AK191933, AK012808, and others.
Gene Ontology Categories:
