siRNA Details
Gene Symbol: Mybpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765246.1 | XP_008763468.1 | 29 | 3414 | ||
XM_008765247.1 | XP_008763469.1 | 29 | 3414 | |||
XM_008765248.1 | XP_008763470.1 | 28 | 3377 | |||
XM_008765249.1 | XP_008763471.1 | 28 | 3370 | |||
XM_008765250.1 | XP_008763472.1 | 28 | 3355 | |||
XM_008765251.1 | XP_008763473.1 | 27 | 3336 | |||
XM_008765252.1 | XP_008763474.1 | 27 | 3333 | |||
XM_008765253.1 | XP_008763475.1 | 28 | 3354 | |||
XM_008765254.1 | XP_008763476.1 | 27 | 3317 | |||
XM_008765255.1 | XP_008763477.1 | 27 | 3336 | |||
XM_008765256.1 | XP_008763478.1 | 26 | 3292 | |||
XM_008765257.1 | XP_008763479.1 | 26 | 3273 | |||
XM_008765258.1 | XP_008763480.1 | 26 | 3272 | |||
GenBank mRNA | FQ214958.1 | - | 4 | 467 | ||
FQ215501.1 | - | 5 | 558 |
