siRNA Details
Gene Symbol: Mbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241463.2 | XP_006241525.1 | 4, 5 | 605 | ||
XM_008765404.1 | XP_008763626.1 | 3, 4 | 296 | |||
XM_008765405.1 | XP_008763627.1 | 4, 5 | 600 | |||
XM_008765406.1 | XP_008763628.1 | 4, 5 | 435 | |||
XM_008765407.1 | XP_008763629.1 | 3, 4 | 296 |
