siRNA Details
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012200.1 | NP_001012200.1 | 7 | 841 | ||
XM_006242102.2 | XP_006242164.1 | 7 | 846 | |||
XM_006242103.2 | XP_006242165.1 | 7 | 892 | |||
XM_006242104.1 | XP_006242166.1 | 6 | 887 | |||
XM_008765764.1 | XP_008763986.1 | 6 | 798 | |||
GenBank mRNA | BC087669.1 | - | 7 | 841 | 7316813 |
