siRNA Details
Gene Symbol: Trmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135876.1 | NP_001129348.1 | 7, 8 | 839 | ||
XM_006242164.2 | XP_006242226.1 | 6, 7 | 924 | |||
XM_008765766.1 | XP_008763988.1 | 6, 7 | 549 | |||
GenBank mRNA | BC161991.1 | - | 6, 7 | 883 | 9032691 |
