siRNA Details
Gene Symbol: Trmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135876.1 | NP_001129348.1 | 9 | 1034 | ||
XM_006242164.2 | XP_006242226.1 | 8 | 1119 | |||
XM_008765766.1 | XP_008763988.1 | 8 | 744 | |||
GenBank mRNA | BC161991.1 | - | 8 | 1078 | 9032691 |
