siRNA Details
Gene Symbol: Dennd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108749.1 | NP_001102219.1 | 20 | 1773 | ||
XM_006242222.2 | XP_006242284.1 | 19 | 3273 | |||
XM_006242224.2 | XP_006242286.1 | 1540 |
