siRNA Details
Gene Symbol: Siae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108759.1 | NP_001102229.1 | 8 | 1149 | ||
GenBank mRNA | BC162050.1 | - | 7 | 1010 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Siae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108759.1 | NP_001102229.1 | 8 | 1149 | ||
GenBank mRNA | BC162050.1 | - | 7 | 1010 |