siRNA Details
Gene Symbol: Dixdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037654.1 | NP_001032743.1 | 3 | 316 | ||
XM_006243042.2 | XP_006243104.1 | 3 | 773 | |||
XM_006243045.2 | XP_006243107.1 | 2 | 128 | |||
XM_008766295.1 | XP_008764517.1 | 3 | 809 | |||
XM_008766297.1 | XP_008764519.1 | 3 | 780 | |||
GenBank mRNA | AY770507.1 | - | 3 | 316 |
