siRNA Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108761.1 | NP_001102231.1 | 18, 19 | 3591 | ||
XM_006243129.2 | XP_006243191.1 | 18, 19 | 3587 | |||
XM_006243131.2 | XP_006243193.1 | 17, 18 | 3374 | |||
XM_008766298.1 | XP_008764520.1 | 17, 18 | 3779 | |||
XM_008766299.1 | XP_008764521.1 | 18, 19 | 3862 | |||
XM_008766300.1 | XP_008764522.1 | 18, 19 | 3371 | |||
XM_008766301.1 | XP_008764523.1 | 17, 18 | 3305 | |||
XM_008766302.1 | XP_008764524.1 | 17, 18 | 4173 |
