siRNA Details
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008380.1 | NP_001008381.1 | 15, 16 | 2698 | ||
XM_006243261.2 | XP_006243323.1 | 15, 16 | 2801 | |||
XM_006243262.2 | XP_006243324.1 | 15, 16 | 2605 | |||
XM_006243263.2 | XP_006243325.1 | 15, 16 | 3130 | |||
GenBank mRNA | BC085932.1 | - | 15, 16 | 2698 | 7112012 | |
FQ216927.1 | - | 5, 6 | 1111 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001001295, NM_172519, NM_001177784, AK029974, BC056939, and others.
Gene Ontology Categories:
