siRNA Details
Gene Symbol: Rsl24d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014212.1 | NP_001014234.1 | 1 | 117 | ||
GenBank mRNA | BC062237.1 | - | 1 | 117 | 6919069 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198609, AK153492, BC089481, AK160194, AK164856.
Gene Ontology Categories:
