siRNA Details
Gene Symbol: Pcbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191883.1 | NP_001178812.1 | 6 | 562 | ||
XM_006243802.2 | XP_006243864.1 | 7 | 1058 | |||
XM_006243803.2 | XP_006243865.1 | 7 | 1057 | |||
GenBank mRNA | AW533143.1 | - | 6 | 492 |
