siRNA Details
Gene Symbol: Fam134a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100760.1 | NP_001094230.1 | 6 | 728 | ||
XM_006245244.2 | XP_006245306.1 | 6 | 750 | |||
GenBank mRNA | BC104673.1 | - | 6 | 538 |
