siRNA Details
Gene Symbol: Atg9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014218.1 | NP_001014240.1 | 8 | 1313 | ||
XM_006245247.2 | XP_006245309.1 | 8 | 1017 | |||
XM_008767262.1 | XP_008765484.1 | 8 | 1680 | |||
XM_008767263.1 | XP_008765485.1 | 8 | 1192 | |||
XM_008767264.1 | XP_008765486.1 | 8 | 1020 | |||
XM_008767265.1 | XP_008765487.1 | 9 | 1078 | |||
XM_008767266.1 | XP_008765488.1 | 9 | 1081 | |||
GenBank mRNA | BC089204.1 | - | 8 | 1313 | 7190326 |
