siRNA Details
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014219.1 | NP_001014241.1 | 10 | 2061 | ||
XM_006245248.2 | XP_006245310.1 | 9 | 1958 | |||
GenBank mRNA | BC081973.1 | - | 10 | 2061 | 7126276 |
