siRNA Details
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108802.1 | NP_001102272.1 | 32, 33 | 7991 | ||
XM_006245252.2 | XP_006245314.1 | 34, 35 | 7953 | |||
XM_006245255.2 | XP_006245317.1 | 27, 28 | 7749 | |||
XM_006245256.2 | XP_006245318.1 | 27, 28 | 5778 | |||
XM_008767267.1 | XP_008765489.1 | 32, 33 | 8053 | |||
XM_008767268.1 | XP_008765490.1 | 32, 33 | 7830 | |||
XM_008767269.1 | XP_008765491.1 | 33, 34 | 8447 |
