siRNA Details
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135893.1 | NP_001129365.1 | 37, 38 | 6706 | ||
NM_001135923.1 | NP_001129395.1 | 36, 37 | 6579 | |||
XM_006256666.2 | XP_006256728.1 | 39, 40 | 6978 | |||
XM_006256667.2 | XP_006256729.1 | 40, 41 | 6986 | |||
XM_006256668.2 | XP_006256730.1 | 39, 40 | 6971 | |||
XM_006256669.2 | XP_006256731.1 | 36, 37 | 6617 | |||
XM_006256670.2 | XP_006256732.1 | 39, 40 | 7222 | |||
XM_006256671.2 | XP_006256733.1 | 36, 37 | 6603 | |||
XM_008773061.1 | XP_008771283.1 | 39, 40 | 7237 |
