siRNA Details
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025747.1 | NP_001020918.1 | 5, 6 | 432 | ||
XM_006257077.2 | XP_006257139.1 | 7, 8 | 589 | |||
GenBank mRNA | BC097987.1 | - | 5, 6 | 432 | 7444864 | |
FQ211792.1 | - | 5, 6 | 517 |
