siRNA Details
Gene Symbol: Hcfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139507.1 | NP_001132979.1 | 7 | 2098 | ||
XM_006229593.2 | XP_006229655.1 | 7 | 2077 | |||
XM_006229594.2 | XP_006229656.1 | 7 | 2076 | |||
XM_006229596.2 | XP_006229658.1 | 7 | 2074 | |||
XM_006229597.1 | XP_006229659.1 | 7 | 2093 |
