siRNA Details
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246362.2 | XP_006246424.1 | 8 | 1156 | ||
XM_006246363.2 | XP_006246425.1 | 8 | 1166 | |||
XM_006246364.2 | XP_006246426.1 | 8 | 1160 | |||
XM_006246365.2 | XP_006246427.1 | 7 | 1063 | |||
XM_008767741.1 | XP_008765963.1 | 8 | 2980 | |||
GenBank mRNA | FQ233211.1 | - | 2 | 123 |
