siRNA Details
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108826.1 | NP_001102296.1 | 13 | 1430 | ||
XM_006246303.2 | XP_006246365.1 | 12 | 1402 | |||
XM_006246304.2 | XP_006246366.1 | 12 | 1378 | |||
XM_006246306.2 | XP_006246368.1 | 12 | 1402 | |||
XM_006246307.2 | XP_006246369.1 | 11 | 1321 | |||
XM_006246310.2 | XP_006246372.1 | 11 | 1226 |
