siRNA Details
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108841.1 | NP_001102311.1 | 704 | |||
XM_008768584.1 | XP_008766806.1 | 3 | 313 | |||
XM_008768585.1 | XP_008766807.1 | 3 | 308 | |||
XM_008768586.1 | XP_008766808.1 | 3 | 308 | |||
XM_008768587.1 | XP_008766809.1 | 3 | 313 |
