siRNA Details
Gene Symbol: Iqcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014230.1 | NP_001014252.1 | 9 | 1004 | ||
XM_006248455.2 | XP_006248517.1 | 9 | 1197 | |||
XM_006248456.2 | XP_006248518.1 | 9 | 1399 | |||
GenBank mRNA | BC087078.1 | - | 9 | 1004 | 7135432 |
