siRNA Details
Gene Symbol: Dzip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252465.2 | XP_006252527.1 | 14 | 1807 | ||
XM_006252466.1 | XP_006252528.1 | 14 | 1680 | |||
XM_008770948.1 | XP_008769170.1 | 13 | 1860 | |||
XM_008770949.1 | XP_008769171.1 | 13 | 1857 |
