siRNA Details
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108896.1 | NP_001102366.1 | 8 | 981 | ||
GenBank mRNA | FQ228966.1 | - | 8 | 899 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108896.1 | NP_001102366.1 | 8 | 981 | ||
GenBank mRNA | FQ228966.1 | - | 8 | 899 |