siRNA Details
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108896.1 | NP_001102366.1 | 6 | 573 | ||
GenBank mRNA | FQ228966.1 | - | 6 | 491 |

More Information
Other Species Targeted

Gene Ontology Categories:
