siRNA Details
Gene Symbol: Dpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008383.1 | NP_001008384.1 | 2 | 451 | ||
XM_006255517.2 | XP_006255579.1 | 3 | 472 | |||
XM_008772521.1 | XP_008770743.1 | 3 | 669 | |||
GenBank mRNA | BC085826.1 | - | 2 | 451 | 7131855 |
