siRNA Details
Gene Symbol: Mical2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139508.1 | NP_001132980.1 | 3, 4 | 597 | ||
XM_006230065.2 | XP_006230127.1 | 5, 6 | 929 | |||
XM_006230066.2 | XP_006230128.1 | 5, 6 | 929 |
