siRNA Details
Gene Symbol: Zbtb24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098667.1 | NP_001092137.1 | 2 | 509 | ||
XM_008773001.1 | XP_008771223.1 | 2 | 2039 | |||
XM_008773002.1 | XP_008771224.1 | 2 | 502 | |||
XM_008773003.1 | XP_008771225.1 | 2 | 2040 |
