siRNA Details
Gene Symbol: Marveld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108936.1 | NP_001102406.1 | 4 | 1449 | ||
XM_008760679.1 | XP_008758901.1 | 4 | 1384 | |||
XM_008760680.1 | XP_008758902.1 | 4 | 1518 | |||
XM_008760681.1 | XP_008758903.1 | 4 | 1486 |
