siRNA Details
Gene Symbol: Skiv2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034093.1 | NP_001029265.1 | 20 | 2473 | ||
GenBank mRNA | BC091305.1 | - | 2 | 114 | ||
CF977889.1 | - | 6 | 589 | |||
CO401332.1 | - | 2 | 113 | |||
CO571909.1 | - | 5 | 450 |
