siRNA Details
Gene Symbol: Hnf4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108939.1 | NP_001102409.1 | 5 | 692 | ||
XM_006232165.2 | XP_006232227.1 | 5 | 1443 | |||
XM_006232166.2 | XP_006232228.1 | 6 | 1286 |
