siRNA Details
Gene Symbol: Slc25a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108947.1 | NP_001102417.1 | 2 | 893 | ||
XM_006232719.2 | XP_006232781.1 | 2 | 728 | |||
XM_006232720.2 | XP_006232782.1 | 2 | 884 | |||
XM_006232721.1 | XP_006232783.1 | 2 | 753 | |||
XM_006232722.1 | XP_006232784.1 | 2 | 729 | |||
XM_006232723.1 | XP_006232785.1 | 2 | 729 |
