siRNA Details
Gene Symbol: Nup210l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191900.1 | NP_001178829.1 | 26, 27 | 3549 | ||
XM_006232724.2 | XP_006232786.1 | 26, 27 | 3606 | |||
XM_008761196.1 | XP_008759418.1 | 26, 27 | 3606 | |||
XM_008761198.1 | XP_008759420.1 | 26, 27 | 3606 | |||
XM_008761199.1 | XP_008759421.1 | 26, 27 | 3606 | |||
XM_008761200.1 | XP_008759422.1 | 26, 27 | 3606 |
