siRNA Details
Gene Symbol: Zbtb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108953.1 | NP_001102423.1 | 2 | 898 | ||
XM_006234101.2 | XP_006234163.1 | 2 | 994 | |||
XM_006234102.1 | XP_006234164.1 | 2 | 1269 |
