siRNA Details
Gene Symbol: Cfl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108982.2 | NP_001102452.2 | 3, 4 | 532 | ||
XM_006240104.2 | XP_006240166.1 | 3, 4 | 619 | |||
GenBank mRNA | FQ216032.1 | - | 1, 2 | 99 | ||
FQ224178.1 | - | 3, 4 | 208 | |||
FQ228820.1 | - | 3, 4 | 535 | |||
FQ228857.1 | - | 3, 4 | 539 |
