siRNA Details
Gene Symbol: NEWGENE_1310847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764977.1 | XP_008763199.1 | 1 | 615 | ||
XM_008764978.1 | XP_008763200.1 | 2 | 784 | |||
XM_008764979.1 | XP_008763201.1 | 2 | 800 | |||
XM_008764980.1 | XP_008763202.1 | 2 | 802 |
Gene Symbol: Bag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008526.2 | NP_001008526.1 | 726 | |||
XM_006240601.2 | XP_006240663.1 | 886 | ||||
XM_006240602.2 | XP_006240664.1 | 826 | ||||
XM_006240603.2 | XP_006240665.1 | 802 | ||||
GenBank mRNA | AY366364.1 | - | 771 | |||
BC088418.1 | - | 726 | 7123242 |
