siRNA Details
Gene Symbol: Caps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108990.1 | NP_001102460.1 | 9 | 847 | ||
XM_006241337.2 | XP_006241399.1 | 19 | 2195 | |||
XM_008765359.1 | XP_008763581.1 | 18 | 1727 |
