siRNA Details
Gene Symbol: Trhde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108991.1 | NP_001102461.1 | 14 | 3300 | ||
XM_008765360.1 | XP_008763582.1 | 10 | 2964 | |||
GenBank mRNA | X80535.1 | - | 13 | 2359 |
