siRNA Details
Gene Symbol: Tbc1d15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241353.2 | XP_006241415.1 | 8 | 966 | ||
XM_345825.7 | XP_345826.3 | 7 | 915 |

More Information
Other Species Targeted

Gene Ontology Categories:
