siRNA Details
Gene Symbol: Tbc1d15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241353.2 | XP_006241415.1 | 15, 16 | 1716 | ||
XM_345825.7 | XP_345826.3 | 14, 15 | 1665 | |||
GenBank mRNA | BC085348.1 | - | 2, 3 | 130 | ||
BC099146.1 | - | 2, 3 | 192 |

More Information
Other Species Targeted

Gene Ontology Categories:
