siRNA Details
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130571.1 | NP_001124043.1 | 26, 27 | 3201 | ||
XM_006241853.2 | XP_006241915.1 | 26, 27 | 3202 | |||
GenBank mRNA | BC168713.1 | - | 26, 27 | 3115 | 8367087 |
