siRNA Details
Gene Symbol: Fam76b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108994.1 | NP_001102464.1 | 4 | 471 | ||
XM_006242526.1 | XP_006242588.1 | 4 | 456 | |||
XM_006242527.1 | XP_006242589.1 | 4 | 456 | |||
XM_006242529.2 | XP_006242591.1 | 4 | 456 | |||
XM_006242530.2 | XP_006242592.1 | 4 | 456 | |||
XM_006242531.2 | XP_006242593.1 | 1 | 20 | |||
GenBank mRNA | FQ212292.1 | - | 4 | 166 |
