siRNA Details
Gene Symbol: Fam76b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108994.1 | NP_001102464.1 | 11 | 1273 | ||
XM_006242529.2 | XP_006242591.1 | 10 | 1200 | |||
XM_006242530.2 | XP_006242592.1 | 10 | 1197 | |||
GenBank mRNA | FQ212292.1 | - | 10 | 907 |
